Skip to Content
MilliporeSigma

257540

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine copper(II)

Dye content ~95 %

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H44CuN4
CAS Number:
Molecular Weight:
596.31
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

composition

Dye content, ~95%

λmax

399 nm

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Cu]n34)c(CC)c5CC

InChI

1S/C36H44N4.Cu/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;/h17-20H,9-16H2,1-8H3;/q-2;+2/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;

InChI key

WYWUPEFEFRBNGN-XTPDIVBZSA-N

General description

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine metalates with copper (II) to form 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine copper(II). CuOEP was used to prepare a mixed adlayer with cobalt phthalocyanine, by immersing an Au(111) substrate.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library