Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

927139

Sigma-Aldrich

DBO

≥95%

Synonym(s):

N,N′-Dibenzyloxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O2
CAS Number:
Molecular Weight:
268.31
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

SMILES string

O=C(C(NCC1=CC=CC=C1)=O)NCC2=CC=CC=C2

InChI

1S/C16H16N2O2/c19-15(17-11-13-7-3-1-4-8-13)16(20)18-12-14-9-5-2-6-10-14/h1-10H,11-12H2,(H,17,19)(H,18,20)

InChI key

AOIKOYRULAZZLJ-UHFFFAOYSA-N

Application

DBO is a ligand for the copper-catalyzed couplings of heteroanilines with (hetero)aryl halides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhixiang Chen et al.
Organic letters, 21(17), 6874-6878 (2019-08-16)
N,N'-Dibenzyloxalamide (DBO) was identified as a powerful ligand for promoting Cu-catalyzed coupling of heteroanilines with (hetero)aryl halides. For (hetero)aryl chlorides, the coupling reaction occurred at 130 °C with 5 mol % CuBr and 10 mol % DBO. For (hetero)aryl bromides/iodides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service