Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

927163

Sigma-Aldrich

BNMO

≥95%

Synonym(s):

N1,N2-Bis(1-naphthalenylmethyl)ethanediamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N2O2
CAS Number:
Molecular Weight:
368.43
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

Application

BNMO is a ligand for the copper-catalyzed coupling of aryl halides and alkyl alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhixiang Chen et al.
Journal of the American Chemical Society, 141(8), 3541-3549 (2019-01-29)
A robust and practical protocol for preparing alkyl aryl ethers has been developed, which relies on using two types of ligands to promote Cu-catalyzed alkoxylation of (hetero)aryl halides. The reaction scope is very general for a variety of coupling partners

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service