Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

924008

Sigma-Aldrich

Silver bromide

≥99.999% trace metals basis

Synonym(s):

Argentous bromide, Silver monobromide, Silver(I) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgBr
CAS Number:
Molecular Weight:
187.77
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes
form:
powder, crystals or chunks

grade

for analytical purposes

Quality Level

Assay

≥99.999% trace metals basis

form

powder, crystals or chunks

mp

432 °C (lit.)

density

6.473 g/mL at 25 °C (lit.)

SMILES string

Br[Ag]

InChI

1S/Ag.BrH/h;1H/q+1;/p-1

InChI key

ADZWSOLPGZMUMY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Silver bromide (AgBr) with 99.999% trace metal purity is an inorganic compound and is used in various fields due to its exceptional light sensitivity, optical properties, and high purity.

Application

Silver bromide (AgBr) is a promising photocatalyst with a narrow bandgap. Three- dimensional hydrogels based with AgBr show remarkable antibacterial properties, making it a viable material for photochromic contact lenses. Graphene-based aerogels as catalysts support for AgBr have also been reported to show high photocatalytic activity, these composites could be used for photodegradation of organic pollutants and have antibacterial applications. AgBr can be utilized in the synthesis of silver bromide nanoparticles, which have applications in catalysis, sensing, and photonic devices. Silver bromide has been used in research areas for creating thin films and coatings with unique optical and electronic properties.

Features and Benefits

Silver bromide with 99.999% trace metal purity minimizes contamination from other metals, making it perfect for high-precision and high-performance applications. This exceptional purity ensures consistent and reliable properties, which are essential for scientific research and advanced industrial processes.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Double Z-scheme system of silver bromide@bismuth tungstate/tungsten trioxide ternary heterojunction with enhanced visible-light photocatalytic activity
Zhang M, et al.
Journal of Colloid and Interface Science, 509, 18-24 (2018)
Fabrication of a three-dimensional porous Z-scheme silver/silver bromide/graphitic carbon nitride@nitrogen-doped graphene aerogel with enhanced visible-light photocatalytic and antibacterial activities
Chen Y, et al.
Journal of Colloid and Interface Science, 536, 389-398 (2019)
Synthesis of a high-performance silver silicate (Ag6Si2O7)/silver bromide (AgBr) photocatalyst with enhanced visible light catalytic activity for refractory organic pollutants
Qin J, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 577, 213-223 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service