Skip to Content
MilliporeSigma
All Photos(1)

Documents

654981

Sigma-Aldrich

Bismuth(III) bromide

anhydrous, powder, 99.998% trace metals basis

Synonym(s):

Bismuth tribromide, Tribromobismuthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiBr3
CAS Number:
Molecular Weight:
448.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.998% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Metal Analysis

density

5.7 g/mL at 25 °C (lit.)

SMILES string

Br[Bi](Br)Br

InChI

1S/Bi.3BrH/h;3*1H/q+3;;;/p-3

InChI key

TXKAQZRUJUNDHI-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyzes the formation of cyclic carbonates, which are important precursors for polycarbonates and other polymeric materials.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service