Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

922269

Sigma-Aldrich

TpOx-Ph-opo-3F

Synonym(s):

2,3,6,11,12-pentakis(pentyloxy)-8-(2,4,6-trifluorophenyl)triphenyleno[1,2-d]oxazole, TpOx-Ph-Fn Series

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H62F3NO6
CAS Number:
Molecular Weight:
830.03
MDL number:
NACRES:
NA.23

description

Polymorph crystalline phase: Solid, Discotic Liquid Crystal, Isotropic

Quality Level

transition temp

crystalline phase to discotic phase 105 °C (heating)
discotic phase to isotropic phase 198 °C (heating)

solubility

soluble (THF > 1 mg/mL, MeCN < 0.07 mg/mL, DMSO < 0.03 mg/mL, DCM > 1 mg /mL
)

λmax

270 nm in ethyl acetate

εmax

103,000 M-1cm-1

fluorescence

λem 503 nm, quantum yield 0.63 (Fluorescence lifetime (ns) = 6.65)

storage temp.

2-8°C

General description

TpOx-Ph-opo-3F is UV excited fluorescent material with a polycyclic aromatic donor-acceptor structure where the triphenoxazole core acts as a donor and the aromatic group on the two position of the oxazole (2,4,6-trifluoro-phenyl) act as the acceptor group. The push-pull, donor-acceptor, structure facilitates intramolecular charge transfer in the excited state that results in a 226 nm emission Stokes Shift. TpOx-Ph-opo-3F is also a photo-conducting Discotic Liquid Crystalline (DLC) material with mesophase transition onset temperature of 105 °C. TpOx-Ph-opo-F is designed for 355 nm and 405 nm excitation with emission at 496 nm with quantum yield of 0.63, high thermal, chemical and photostability. TpOx-Ph-opo-3F has potential uses in fluorescent dye staining, organic electronic and photonics, and imaging applications.

Application

Fluorescent materials have a range of unique properties that allows for potential use in a range of applications:

  • Fluorescent dye staining
UV fluorescent materials can be used to dope polymer and resin composites, including 3D applications, and further identified using fluorescence emission.

  • Organic semiconductor for organic electronic and photonic applications
These organic discotic liquid crystals display columnar hexagonal mesophase, and were incorporated into OPV device as a hole transport layer and donor material. Similar compounds have been used in organic light emitting devices (OLEDs), organic photovoltaic device (OPVs) and organic field-effect transistor (OFETs) as hole transport material.

  • Multi-photon microscopy
These UV fluorescent materials have been used in multi-photon microscopy, showing remarkable image resolution relative to commercial fluorophores in the multiphoton microscopy imaging. By perfusing it into an ex-planted human liver, it was possible to observe fluorophore staining of the hepatocytes and T-cells in the liver vasculature. During the live liver imaging these probes have been shown to perfuse effectively in human tissue and emit brightly without significant cytotoxicity during the course of the experiment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Triphenoxazoles exhibit tunable properties, liquid crystallinity, and potential for modification.

Triphenoxazoles exhibit tunable properties, liquid crystallinity, and potential for modification.

Triphenoxazoles exhibit tunable properties, liquid crystallinity, and potential for modification.

Triphenoxazoles exhibit tunable properties, liquid crystallinity, and potential for modification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service