Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775665

Sigma-Aldrich

2,3,4,6-Tetra-O-acetyl-D-galactopyranose

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O10
CAS Number:
Molecular Weight:
348.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

bp

107-110 °C (lit.)

storage temp.

2-8°C

SMILES string

OC1O[C@H](COC(C)=O)[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O

InChI

1S/C14H20O10/c1-6(15)20-5-10-11(21-7(2)16)12(22-8(3)17)13(14(19)24-10)23-9(4)18/h10-14,19H,5H2,1-4H3/t10-,11+,12+,13-,14?/m1/s1

InChI key

IEOLRPPTIGNUNP-RRYROLNDSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service