Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

920959

Sigma-Aldrich

Graphene, monolayer film

on Si/SiO2 wafer, diam. 100 mm (4 in.)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
rGO
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Product Name

Monolayer Graphene on Si/SiO2 wafer, diam. 100 mm (4 in.)

description

Graphene Coverage: 100% with sporadic adlayers
Metal Impurities: 1.00e10 – 5.00e10 (at/cm) substrate
Monolayer Graphene Transparency: >97%
Raman D/G ratio: Indistinguishable to 0.03
Substrate Type/Doping: P/B
430 Ω/sq ±50 Ω/sq

Quality Level

resistivity

1-10 Ω-cm

diam.

100 mm (4 in.)

grain size

>40 μm

semiconductor properties

(mobility>2700 cm2/V·s) (FET mobility)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

General description

Each graphene unit sold is accompanied by a specification sheet unique to the S/N and contains batch-specific metrics. This eliminates guesswork and ensures that you only get the best, well-characterized, and QA-passed graphene.

Application

Graphene is one of the most promising carbon nanomaterials, and it was first isolated in 2004 by Novoselov, Geim et al, leading to a Nobel prize. It consists of a two-dimensional single layer of sp2 hybridized carbon with hexagonal lattice. It has attracted significant attention in the field of high-performance electronic devices because of its excellent electronic and chemical properties, and large specific surface area . Among the main potential applications of our monolayer graphene on Si/SiO2 wafers are transistors and sensors.

Storage and Stability

To ensure the maximum shelf life of your graphene sample, especially if it rests on copper foil, it is best stored under vacuum or in inert atmosphere (Argon or Nitrogen) conditions once the vacuum sealed package has been opened.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graphene-Based Transparent Conductive Electrodes
Yu K, et al.
Science null
The electronic properties of graphene
Neto A.C, et al.
Reviews of Modern Physics, 81 (1), 109-162 (2009)
Uptake of H2 and CO2 by graphene
Ghosh A, et al.
The Journal of Physical Chemistry, 112 (40), 15704-15707 (2008)
Electric field effect in atomically thin carbon films
Novoselov K S, et al.
Science, 306 (5696), 666-669 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service