Skip to Content
MilliporeSigma
All Photos(2)

Documents

777684

Sigma-Aldrich

Reduced graphene oxide

Synonym(s):

rGO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxOyHz
UNSPSC Code:
12352103
NACRES:
NA.23

description

chemically reduced

form

powder

composition

Carbon, ≥75 wt. %
Nitrogen, >5 wt. %
Oxygen, <22 wt. %

surface area

450 m2/g

conductivity

>600 S/m

solubility

water: insoluble

Looking for similar products? Visit Product Comparison Guide

General description

The structure of graphene oxide is similar to graphene containing residual oxygen, heteroatoms and structural defects. Reduced graphene oxide (rGO) can be extracted as thin film from an aqueous solution of graphite oxide (GO) slurry. Many modern procedures for the synthesis of GO are based on the Hummers method, in which graphite is oxidized by a solution of potassium permanganate in sulfuric acid. Reduction of GO using hydrazine has been reported. However, hydrazine is highly toxic and can potentially functionalize GO with nitrogen heteroatoms. Because of these issues, alternatives to hydrazine including NaBH4, ascorbic acid, and HI, among others have been used for the reduction of GO. GO can be reduced as a thin film or in an aqueous solution. Reduction methods have been recently reviewed.

Application

The spin-coated reduced graphene oxide (rGO) thin films have been used as annode buffer layer to fabricate bulk heterojunction org. solar cells. PEGylated rGO may be used as:
  • chemical sensors, biosensors
  • as transparent electrodes in light emitting diodes (LEDs) and ITOs
  • in lithium ion batteries.

Preparation Note

  • Reduction method: Chemically reduced
  • Color: Black
  • Odor: Odorless
  • Solubility: Insoluble
  • Dispersability: It can be dispersed at low concentrations (<0.1mg/mL) in NPM, DMSO, DMF
  • Electrical conductivity: > 600S/m
  • BET surface area: 450 m2/g

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Somayeh Mousavi Nodoushan et al.
Biosensors & bioelectronics, 127, 221-228 (2019-01-10)
Detection of staphylococcal enterotoxin B (SEB) as a bacterial toxin causing severe food poisoning is of great importance. Herein, we developed an electrochemical aptasensor for SEB detection using a screen printed electrode modified with reduced graphene oxide (rGO) and gold
Goki Eda et al.
Nature nanotechnology, 3(5), 270-274 (2008-07-26)
The integration of novel materials such as single-walled carbon nanotubes and nanowires into devices has been challenging, but developments in transfer printing and solution-based methods now allow these materials to be incorporated into large-area electronics. Similar efforts are now being
Monique C P Mendonça et al.
Nanomaterials (Basel, Switzerland), 9(6) (2019-06-15)
Graphene-based nanomaterials (GBNs) possess unique physicochemical properties, allowing a wide range of applications in physical, chemical, and biomedical fields. Although GBNs are broadly used, information about their adverse effects on ecosystem health, especially in the terrestrial environment, is limited. Therefore
Zhang; Liming;
Journal of Material Chemistry B: Materials for Biology and Medicine, 1(6), 749-755 (2013)
Rosa Garriga et al.
Nanomaterials (Basel, Switzerland), 10(8) (2020-08-23)
Carbon nanomaterials have attracted increasing attention in biomedicine recently to be used as drug nanocarriers suitable for medical treatments, due to their large surface area, high cellular internalization and preferential tumor accumulation, that enable these nanomaterials to transport chemotherapeutic agents

Articles

Graphene oxide, a monomolecular layer of graphite with oxygen functionalities, holds unique properties valuable for various applications in materials science.

Graphene oxide, a monomolecular layer of graphite with oxygen functionalities, holds unique properties valuable for various applications in materials science.

Graphene oxide, a monomolecular layer of graphite with oxygen functionalities, holds unique properties valuable for various applications in materials science.

Graphene oxide, a monomolecular layer of graphite with oxygen functionalities, holds unique properties valuable for various applications in materials science.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service