Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

916803

Sigma-Aldrich

1 M Biphenyl in THF

a precursor solution to prepare the prelithiation/presodiation reagent for disordered carbon, SiO, Sn, Sb, P, and S electrodes

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352401
NACRES:
NA.23

Quality Level

form

liquid

concentration

1 M

impurities

≤1000 ppm H2O

application(s)

battery manufacturing

storage temp.

2-8°C

SMILES string

c2(ccccc2)c1ccccc1

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Application

This product is biphenyl (Bp) solution in anhydrous THF (1 mol/L) is a precursor reagent that could be easily handled to prepare prelithiation or presodiation reagent. By direct reaction of Li metal, it forms a strong reducing reagent of Li-Bp. The resulting Li-Bp/THF reagent has the redox potential of 0.41 V vs. Li/Li+, which is applicable to prelithiate most of the active materials for Li ion batteries, such as Sn, Sb, P, S, SiO and other metal oxide. On the other hand, a presodiation reagent can be prepared by reacting Bp/THF solution with Na metal. Na-Bp/THF reagent can be used to presodiate hard carbon, a promising anode material in Na ion batteries.

Preparation Note

Suggested Directions for use:
  • Step 1 Preparation of Prelithiation/presodiation reagent
Before use of Li metal, scrub the lithium surface to remove oxide and nitride layers.
To prepare 1 M Li-Bp/THF reagent, > 0.07 g of Li metal is added into every 10 ml of 1 M BP/THF solution
To prepare 1 M Na-Bp/THF reagent, > 0.23 g of Na metal is added into every 10 ml of 1 M BP/THF solution
  • Step 2 Prelithiation/presodiation of electrodes
For a 1 cm diameter electrode with 1-3 mg acitve material per cm2, soak (10-60 min, times will vary with material) the electrode in ∼0.5-1 mL of prelithiation/presodiation reagent. Discard the solution and then wash the electrode twice with THF. Longer prelithiation/presodiation time may cause electrode structure deformation, leading to a decreased discharge/charge capacity.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gongwei Wang et al.
ACS applied materials & interfaces, 11(9), 8699-8703 (2019-02-20)
This study reports an ambient-air-tolerant approach for negative electrode prelithiation by using 1 M lithium-biphenyl (Li-Bp)/tetrahydrofuran (THF) solution as the prelithiation reagent. Key to this strategy are the relatively stable nature of 1 M Li-Bp/THF in ambient air and the
High performance lithium-ion and lithium−sulfur batteries using prelithiated phosphorus/carbon composite anode..
Wang F, et al.
Energy Storage Materials, 24, 147-152 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service