Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

921106

Sigma-Aldrich

Lithium metal scrubbing solution for battery testing Naphthalene

0.1 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8
CAS Number:
Molecular Weight:
128.17
UNSPSC Code:
12352401
NACRES:
NA.23

form

solution

Quality Level

concentration

0.1 M in THF

application(s)

battery manufacturing

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

Application

This is a chemical polishing reagent that can smooth the surface of lithium metal and remove native surface impurities. Treatment of lithium metal with this solution has been shown to improve the cycle life and rate performance of lithium metal batteries. Use of this reagent to treat your lithium foil removes the need for scraping to give you a clean lithium surface and may increase the quality and useful testing time for your half-cells.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-1.3 °F

Flash Point(C)

-18.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemically polished lithium metal anode for high energy lithium metal batteries.
Tang W, et al.
Energy Storage Materials, 14, 289-289 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service