Skip to Content
MilliporeSigma
All Photos(1)

Documents

916447

Sigma-Aldrich

Thallium(I) iodide

AnhydroBeads, ~10 mesh, 99.999% trace metals basis

Synonym(s):

Thallium iodide, Thallium monoiodide, Thallium(1+) iodide, Thallous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlI
CAS Number:
Molecular Weight:
331.29
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

powder

particle size

~10 mesh

mp

440 °C (lit.)

density

7.29 g/mL at 25 °C (lit.)

storage temp.

10-30°C

SMILES string

I[Tl]

InChI

1S/HI.Tl/h1H;/q;+1/p-1

InChI key

CMJCEVKJYRZMIA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Thallium iodide is used for synthesis of thallium chalcohalides for X-ray and γ-ray detection and synthesis of thallium cadmium iodide nanostructures for its photocatalytic properties.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple synthesis-controlled fabrication of thallium cadmium iodide nanostructures via a novel route and photocatalytic investigation in degradation of toxic dyes
Ghanbari M, et al.
Inorgorganica Chimica Acta, 455(1), 88-97 (2017)
Thallium Chalcohalides for X-ray and γ-ray Detection
Johnsen S, et al.
Journal of the American Chemical Society, 133(26), 10030-10033 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service