Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

916129

Sigma-Aldrich

1,10-Phenanthroline nickel (ll) dichloride

Synonym(s):

phenNiCl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2N2Ni
CAS Number:
Molecular Weight:
309.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

powder or crystals

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

[Ni].Cl.Cl.n1c2c3ncccc3ccc2ccc1

InChI

1S/C12H8N2.2ClH.Ni/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;;/h1-8H;2*1H;

InChI key

CHVPLVVTLIMTJX-UHFFFAOYSA-N

Application

1,10-Phenanthroline nickel (ll) dichloride is a Ni precatalyst used for a variety of cross-coupling reactions, including N-alkylation of amides and arylation of alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng-Jun Wang et al.
Organic letters, 21(16), 6543-6547 (2019-08-08)
A radical-mediated functionalization of allyl alcohol derived partners with a variety of alkyl 1,4-dihydropyridines via photoredox/nickel dual catalysis is described. This transformation transpires with high linear and E-selectivity, avoiding the requirement of harsh conditions (e.g., strong base, elevated temperature). Additionally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service