Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

908118

Sigma-Aldrich

Palladium on carbon

Evonik Noblyst® P1145 5% Pd

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder

contains

~50% water

extent of labeling

5 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

activated carbon support

SMILES string

[Pd]

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Evonik Noblyst® P1145 5% Pd is a palladium on carbon catalyst. It has been shown to be effective in the following reactions:
  • hydrogenation aliphatic nitro groups
  • hydrogenolysis reactions
  • hydrogenation of aldehydes and ketones
  • dehydrogenation of carbocyclic rings to aromatics
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Other Notes

Unit weight is of net dry catalyst.

Legal Information

Manufactured by Evonik Degussa GmbH.
Noblyst is a registered trademark of Evonik Degussa GmbH

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service