Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

907502

Sigma-Aldrich

Potassium 5-bromo-1H-indole-1-carbodithioate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5BrKNS2
Molecular Weight:
310.28
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

179-185 °C

storage temp.

2-8°C

Application

Potassium 5-bromo-1H-indole-1-carbodithioate is an organic photocatalyst shown to activate alkyl electrophiles via an SN2 pathway.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bertrand Schweitzer-Chaput et al.
Nature chemistry, 11(2), 129-135 (2018-12-05)
Chemists extensively use free radical reactivity for applications in organic synthesis, materials science, and life science. Traditionally, generating radicals requires strategies that exploit the bond dissociation energy or the redox properties of the precursors. Here, we disclose a photochemical catalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service