Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

906298

Sigma-Aldrich

3-(4-Bromophenyl)-3-(trifluoromethyl)-3H-diazirine

Synonym(s):

Probe building block for photoaffinity labeling, Suzuki-Miyaura-compatible multifunctional photo probe building block, Trifluoromethyl diazirine aryl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4BrF3N2
CAS Number:
Molecular Weight:
265.03
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

liquid

reaction suitability

reagent type: cross-linking reagent

storage temp.

2-8°C

SMILES string

BrC1=CC=C(C2(N=N2)C(F)(F)F)C=C1

InChI

1S/C8H4BrF3N2/c9-6-3-1-5(2-4-6)7(13-14-7)8(10,11)12/h1-4H

InChI key

YBEXZCGWGWNLQQ-UHFFFAOYSA-N

Application

3-(4-Bromophenyl)-3-(trifluoromethyl)-3H-diazirine is a Suzuki-Miyaura (S−M)-compatible building block tailored for the design of photoaffinity probes (PAPs). Photoaffinity labeling (PAL) is of growing interest in chemical biology and drug discovery research for use in protein profiling; covalent inhibitors; protein-protein interactions and complexes; and target engagement, identification, or validation studies. Diazirines are desirable photoactivable groups for chemical probes; however, their installation is not always straightforward. In response, diazirine-tolerant S-M coupling conditions were developed by Ichiishi, et al, using 3-(4-bromophenyl)-3-(trifluoromethyl)-3H-diazirine and shown to be efficient and scalable with minimal perturbation of the diazirine.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoko Ichiishi et al.
ACS medicinal chemistry letters, 10(1), 56-61 (2019-01-19)
Access to high quality photoaffinity probe molecules is often constrained by synthetic limitations related to diazirine installation. A survey of recently published photoaffinity probe syntheses identified the Suzuki-Miyaura (S-M) coupling reaction, ubiquitous in drug discovery, as being underutilized to incorporate
Formate assay in body fluids: application in methanol poisoning.
A B Makar et al.
Biochemical medicine, 13(2), 117-126 (1975-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service