Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900831

Sigma-Aldrich

3-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)propanoic acid

Synonym(s):

Carboxyl diazirine alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O2
CAS Number:
Molecular Weight:
166.18
MDL number:
UNSPSC Code:
12352106

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry

availability

available only in USA

storage temp.

−20°C

SMILES string

C#CCCC1(N=N1)CCC(O)=O

InChI

1S/C8H10N2O2/c1-2-3-5-8(9-10-8)6-4-7(11)12/h1H,3-6H2,(H,11,12)

InChI key

JRNSKSYASHYNCM-UHFFFAOYSA-N

Application

Used for chemical probe synthesis, this trifunctional building block contains a light-activated diazirine, alkyne tag, and carboxylic acid synthetic handle. When appended to a ligand or pharmacophore through its acid linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service