Skip to Content
MilliporeSigma
All Photos(1)

Documents

903981

Sigma-Aldrich

2,3,7,8-Tetrafluorothianthrene-S-oxide

≥95%

Synonym(s):

2,3,7,8-Tetrafluorothianthrene-5-oxide, Fluorinated sulfoxide-based thianthrene reagent, Ritter C-H functionalization linchpin, TFT S-oxide, TFT-reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4F4OS2
CAS Number:
Molecular Weight:
304.28
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

solid

mp

253 °C

storage temp.

−20°C

Application

2,3,7,8-Tetrafluorothianthrene-S-oxide (TFT S-oxide) is a fluorinated sulfoxide-based thianthrene reagent for late-stage C-H functionalization. Developed by the Ritter Lab, C-H functionalization by thianthrenation with TFT S-oxide proceeds with >99% selectivity and furnishes functionalized arenes serving as synthetic linchpins for further participation in diverse transformations.
The nonfluorinated thianthrene S-oxide is also available as catalog# 903973 for all arenes more electron-rich than anisole.

Legal Information

Patent Application EP18204755.5
This product is manufactured pursuant to a license with Studiengesellschaft Kohle mbH

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Berger et al.
Nature, 567(7747), 223-228 (2019-03-15)
Direct C-H functionalization can quickly increase useful structural and functional molecular complexity1-3. Site selectivity can sometimes be achieved through appropriate directing groups or substitution patterns1-4-in the absence of such functionality, most aromatic C-H functionalization reactions provide more than one product

Related Content

The Ritter lab currently focuses on fluorination chemistry for late-stage functionalization of complex natural and unnatural products. PhenoFluor™ has been developed as a general reagent for the selective, predictable, direct deoxyfluorination of complex alcohols and phenols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service