Skip to Content
MilliporeSigma
All Photos(1)

Documents

922560

Sigma-Aldrich

S-(Trifluoromethyl)thianthrenium triflate

≥95%

Synonym(s):

TT-CF3+ OTf-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8F6O3S3
Molecular Weight:
434.40
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

145-148 °C

Application

S-(Trifluoromethyl)thianthrenium triflate (TT-CF3+ OTf-) is a trifluoromethylation reagent active for a diverse substrate range. Developed by the Ritter Lab, the enchanced activity is accomplished by participation in either nucleophilic, radical, or electrophilic mechanisms depending on the selected reaction conditions, allowing for the introduction of trifluoromethyl moieties to many substrates with a high functional group tolerance.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Patent application: EP21169034.2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Jia et al.
Journal of the American Chemical Society, 143(20), 7623-7628 (2021-05-15)
Here we report the synthesis and application of trifluoromethyl thianthrenium triflate (TT-CF3+OTf-) as a novel trifluoromethylating reagent, which is conveniently accessible in a single step from thianthrene and triflic anhydride. We demonstrate the use of TT-CF3+OTf- in electrophilic, radical, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service