Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901747

Sigma-Aldrich

7-(3-Bromo-4-methoxybenzyl)-1-ethyl-8-(((1R,2R)-2-hydroxycyclopentyl)amino)-3-(2-hydroxyethyl)-3,7-dihydro-1H-purine-2,6-dione

Synonym(s):

Aryl halide chemistry informer library compound X12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28BrN5O5
CAS Number:
Molecular Weight:
522.39
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

form

powder or crystals

SMILES string

Brc1c(ccc(c1)C[n]2c3c(nc2NC4C(CCC4)O)N(C(=O)N(C3=O)CC)CCO)OC

InChI

1S/C22H28BrN5O5/c1-3-26-20(31)18-19(27(9-10-29)22(26)32)25-21(24-15-5-4-6-16(15)30)28(18)12-13-7-8-17(33-2)14(23)11-13/h7-8,11,15-16,29-30H,3-6,9-10,12H2,1-2H3,(H,24,25)

InChI key

RCJYGWGQCPDYSL-UHFFFAOYSA-N

Application

This product is Informer compound X12 of the Aryl Halide Chemistry Informer Library developed by chemists at Merck & Co., Inc., Kenilworth, NJ, U.S.., Inc., Kenilworth, NJ, U.S., which contains 18 drug-like molecules representative of those encountered in complex synthesis. By screening a new reaction against the Informer Library, chemists can directly compare and analyze a reactions′s successes and shortcomings among different methods and various research teams. It may also be used to faciliate deeper method development for performance or utility.

Caution

Not Fully Tested.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Buitrago Santanilla et al.
Science (New York, N.Y.), 347(6217), 49-53 (2015-01-03)
At the forefront of new synthetic endeavors, such as drug discovery or natural product synthesis, large quantities of material are rarely available and timelines are tight. A miniaturized automation platform enabling high-throughput experimentation for synthetic route scouting to identify conditions
Peter S Kutchukian et al.
Chemical science, 7(4), 2604-2613 (2016-04-21)
Major new advances in synthetic chemistry methods are typically reported using simple, non-standardized reaction substrates, and reaction failures are rarely documented. This makes the evaluation and choice of a synthetic method difficult. We report a standardized complex molecule diagnostic approach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service