Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

804967

Sigma-Aldrich

rac-BINAP-Pd-G3

95%

Synonym(s):

[2′-(amino-κN)[1,1′-biphenyl]-2-yl-κC][[2′-(diphenylphosphino)[1,1′-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)- palladium 

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H45NO3P2PdS
CAS Number:
Molecular Weight:
992.40
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

175-180 °C

functional group

phosphine

SMILES string

CS(O[Pd-2]1([P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=C(C5=C6C(C=CC=C6)=CC=C5P(C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9C=C4)C%10=C(C%11=C([NH2+]1)C=CC=C%11)C=CC=C%10)(=O)=O

InChI

1S/C44H32P2.C12H10N.CH4O3S.Pd/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h1-32H;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

BVYJEOTVUJMCBM-UHFFFAOYSA-M

General description

rac-BINAP-Pd-G3 is a third generation (G3) Buchwald precatalyst. t is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. rac-BINAP-Pd-G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio. It

Application

Pd precatalyst for cross-coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

See All

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service