Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901627

Sigma-Aldrich

Pym-DATB Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H20B4N6O3
CAS Number:
Molecular Weight:
579.78
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder or crystals

reaction suitability

core: boron
reagent type: catalyst

SMILES string

B21N3B(OB(O2)c7c(cccc7)c8ncnc9c8NB(c%10c9cccc%10)O)c4c(cccc4)c5ncnc(c53)c6c1cccc6

InChI

1S/C32H20B4N6O3/c43-33-23-13-5-1-9-19(23)27-31(41-33)28(38-17-37-27)22-12-4-8-16-26(22)36-44-34-24-14-6-2-10-20(24)29-32-30(40-18-39-29)21-11-3-7-15-25(21)35(45-36)42(32)34/h1-18,41,43H

InChI key

UJSSLUDYAHIVLK-UHFFFAOYSA-N

Application

Pym-DATB catalyst is a bench-stable pyrimidine derivative of the B3NO2 six-membered heterocycle 1,3-dioxa-5-aza-2,4,6-triborinane (DATB). Researchers demonstrated Pym-DATB to be an efficient catalyst for dehydrative direct amidation of carboxylic acids and amines to produce a diverse amide products.


Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher R Opie et al.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-02-17)
The B3 NO2 six-membered heterocycle (1,3-dioxa-5-aza-2,4,6-triborinane=DATB), comprising three different non-carbon period 2 elements, has been recently demonstrated to be a powerful catalyst for dehydrative condensation of carboxylic acids and amines. The tedious synthesis of DATB, however, has significantly diminished its utility

Related Content

Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.

Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.

Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.

Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service