909254
2,7-Difluoro-9-mesityl-10-methylacridinium tetrafluoroborate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C23H20BF6N
Molecular Weight:
435.21
UNSPSC Code:
12352101
NACRES:
NA.22
form
powder or crystals
reaction suitability
reaction type: Photocatalysis
reagent type: catalyst
Related Categories
Application
Robust acridinium-based photocatalyst for use as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service