Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900452

Sigma-Aldrich

Graphene paper

size 11.5 in. × 23.5 in., thickness 240 μm

Synonym(s):

Graphene foil, XG Leaf® B 240 μ

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12141908
NACRES:
NA.23

form

sheet

Quality Level

size

11.5 in. × 23.5 in.

thickness

240 μm

density

1.9 g/cm3

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

XG Leaf® is a more effective solution than aluminum and copper foils for a wide variety of applications to optimize thermal conductivity, thermal spreading, electrical properties, and corrosion resistance. XG Leaf® is a thin, flexible, and lightweight sheet product built on a foundation of XG Sciences, xGnP® graphene nanoplatelets. By precisely tailoring the composition, density, and our proprietary manufacturing process materials are created with unique properties to optimize thermal and electrical conductivity. Different types of XG Leaf® graphene paper offer outstanding thermal and electrical properties.
  • Thermal conductivity and spreading: in-plane conductivity above 550 W/mK.
  • Electrical properties: formulations are available with surface resistivity ranging as low as 0.04 Ω/sq.
Potential applications include:
  • Heat dissipation.
  • Resistive heating.
  • EMI shielding.
  • Electrodes (thin battery, super capacitor, Li-air etc).
  • Barrier.
  • Layering in composites.
  • Sensors.

Physical properties

  • Thermal conductivity: 550 W/mK (in-plane), 3 W/mK (through plane).
  • Electrical conductivity: 3700 S/cm (in-plane).
  • Electrical resistivity: surface 0.06 Ω/sq, sheet 2.7 μΩ-m.
  • EMI shielding: 53 dB @ 30 MHz, 58 dB @ 1.5 GHz.

Legal Information

XG Leaf is a registered trademark of XG Sciences, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Advances in scalable synthesis and processing of two-dimensional materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service