Skip to Content
MilliporeSigma
All Photos(1)

Documents

752584

Sigma-Aldrich

Gold nanoparticles

10 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free

Synonym(s):

Au NP, Gold colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles
suspension

concentration

~6.0E+12 particles/mL

OD

1

diameter

10 nm

λmax

515-519 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The gold nanoparticles with a 10 nm diameter and stabilized suspension in 0.1 mM PBS are versatile material with unique properties that make them suitable for various applications. These nanoparticles exhibit exceptional thermo-mechanical properties, a high surface area, and low toxicity, contributing to their wide spread utility. The potential applications of these gold nanoparticles include neutrophil labeling for monitoring the impact of various drugs, immobilization of proteins in biosensor applications, and serving as a nano-carrier in drug delivery systems for the controlled release of hydrophobic drugs.

Application

  • Development of a biophotonic fiber sensor using direct-taper and anti-taper techniques with seven-core and four-core fiber for the detection of doxorubicin in cancer treatment.: Gold nanoparticles play a crucial role in enhancing the sensitivity and precision of fiber optic sensors for detecting chemotherapy drugs, demonstrating their significant potential in medical diagnostics and pharmaceutical research (Li et al., 2024).
  • Generalized ratiometric surface-enhanced Raman scattering biosensor for okadaic acid in food based on Au-triggered signal amplification.: Gold nanoparticles are utilized to amplify Raman scattering signals, improving the detection of toxins in food products, which is essential for food safety and public health research (Wei et al., 2024).

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Matsunaga et al.
Chemosphere, 224, 538-543 (2019-03-06)
A simple analytical method was developed to determine the arsenite (As(III)) concentration using a DNA aptamer and gold nanoparticles (AuNPs). Prior to sample measurements, the method sensing mechanism was confirmed by analyzing the particle size of the AuNPs at each
Emir Yasun et al.
Analytica chimica acta, 1106, 79-87 (2020-03-09)
Rapid and accurate biosensing with low concentrations of the analytes is usually challenged by the diffusion limited reaction kinetics. Thus, as a remedy, long incubation times or excess amounts of the reagents are employed to ensure the reactions to go

Articles

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Sustainable energy sources with high production efficiency are crucial for meeting increasing energy demand.

Gold (Au) nanoparticles have tunable optical and electronic properties and are used in a number of applications including photovoltaics, sensors, drug delivery & catalysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service