Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900074

Sigma-Aldrich

Vitamin K2 (MK-7)-(5,6,7,8-d4,2-methyl-d3)

≥98 atom % D, ≥95% (CP)

Synonym(s):

Menaquinone-7-(5,6,7,8-d4,2-methyl-d3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46D7H57O2
Molecular Weight:
656.04
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

yellow

mp

54 °C

mass shift

M+7

storage temp.

−20°C

SMILES string

O=C1C(C([2H])([2H])[2H])=C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)C(C2=C([2H])C([2H])=C([2H])C([2H])=C21)=O

InChI

1S/C46H64O2/c1-34(2)18-12-19-35(3)20-13-21-36(4)22-14-23-37(5)24-15-25-38(6)26-16-27-39(7)28-17-29-40(8)32-33-42-41(9)45(47)43-30-10-11-31-44(43)46(42)48/h10-11,18,20,22,24,26,28,30-32H,12-17,19,21,23,25,27,29,33H2,1-9H3/b35-20+,36-22+,37-24+,38-26+,39-28+,40-32+/i9D3,10D,11D,30D,31D

InChI key

RAKQPZMEYJZGPI-HRQALKRPSA-N

General description

Vitamin K2, also known as menaquinone, is a polyisoprenoid-substituted napthoquinone. Menaquinone-7 (MK-7) is a subtype of vitamin K2. It is found in animals, soy protein, fermented products and is primarily produced by bacteria.
Vitamin K2 (MK-7)-(5,6,7,8-d4,2-methyl-d3) is a deuterated vitamin K2 wherein C-5, C-6, C-7, C-8, and 2-methyl protons are replaced by deuterium.

Application

Vitamin K2 (MK-7)-(5,6,7,8-d4,2-methyl-d3) can be used as a stable isotope internal standard for data interpretation in specific biological samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of menaquinone (vitamin K2)-7 by Bacillus subtilis
Sato T, et al.
Journal of Bioscience and Bioengineering, 91(1), 16-20 (2001)
Vitamin K
Textbook of Veterinary Physiological Chemistry (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service