Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88719

Sigma-Aldrich

(R)-1-[(SP)-2-(Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine

≥97%

Synonym(s):

(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS), Josiphos SL-J002-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40FeP2
CAS Number:
Molecular Weight:
542.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

crystals

optical purity

ee: ≥99%

functional group

phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)P(C(C)(C)C)C(C)(C)C

InChI

1S/C27H35P2.C5H5.Fe/c1-21(29(26(2,3)4)27(5,6)7)24-19-14-20-25(24)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;1-2-4-5-3-1;/h8-21H,1-7H3;1-5H;/t21-;;/m1../s1

InChI key

STAGMXYQPGLLTD-GHVWMZMZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-1-[(SP)-2-(Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine (PPF-tBu) is a chiral ferrocenyl diphosphine ligand.
sold in collaboration with Solvias AG

Application

It may be used as a ligand in the palladium-catalyzed alkoxycarbonylation of 6-bromo-1,4-benzodioxane with sodium tert-butoxide to form the corresponding tert-butyl ester.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An Efficient Method for the Preparation of Tertiary Esters by Palladium-Catalyzed Alkoxycarbonylation of Aryl Bromides.
Xin, Zhuo, et al.
Organic Letters, 14(1), 284-287 (2011)
Privileged ligands
Aldrich Chemfiles, 6(8), 12-12 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service