Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861545

Sigma-Aldrich

N10-(Trifluoroacetyl)pteroic acid

95%

Synonym(s):

4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl](2,2,2-trifluoroacetyl)amino]benzoic acid, N10-Trifluoroacetylpteroic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11F3N6O4
CAS Number:
Molecular Weight:
408.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

270 °C (dec.) (lit.)

functional group

amide
carboxylic acid
fluoro

SMILES string

Nc1nc(O)c2nc(CN(c3ccc(cc3)C(O)=O)C(=O)C(F)(F)F)cnc2n1

InChI

1S/C16H11F3N6O4/c17-16(18,19)14(29)25(9-3-1-7(2-4-9)13(27)28)6-8-5-21-11-10(22-8)12(26)24-15(20)23-11/h1-5H,6H2,(H,27,28)(H3,20,21,23,24,26)

InChI key

IJGIHDXKYQLIMA-UHFFFAOYSA-N

Application

N10-(Trifluoroacetyl)pteroic acid is a structural component of folic acid. It can be used as a precursor to synthesize folic acid derivatives, γ-azido modified folic acid, and pteroyl-γ-glutamate-cysteine.

Other Notes

Remainder DMF

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pteroyl-γ-glutamate-cysteine synthesis and its application in folate receptor-mediated cancer cell targeting using folate-tethered liposomes.
Zhang Y, et al.
Analytical Biochemistry, 332(1), 168-177 (2004)
The facile synthesis of multifunctional PAMAM dendrimer conjugates through copper-free click chemistry.
Huang B, et al.
Bioorganic & Medicinal Chemistry Letters, 22(9), 3152-3156 (2012)
Self?Assembly of Folic Acid Derivatives: Induction of Supramolecular Chirality by Hierarchical Chiral Structures.
Kamikawa Y, et al.
Chemistry?A European Journal , 10(23), 5942-5951 (2004)
Chun-Yen Ke et al.
Journal of the American Chemical Society, 127(20), 7421-7426 (2005-05-19)
The cell membrane folate receptor is a potential molecular target for tumor-selective drug delivery. To probe structural requirements for folate receptor targeting with low molecular weight radiometal chelates, specifically the role of the amino acid fragment of folic acid (pteroylglutamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service