Skip to Content
MilliporeSigma
All Photos(1)

Documents

83901

Sigma-Aldrich

Rink amide 4-methylbenzhydrylamine, polymer-bound

extent of labeling: ~1.1 mmol/g loading

Synonym(s):

4-(2′,4′-Dimethoxyphenyl-Fmoc-aminmethyl)-phenoxyacetamido-methylbenzhydryl amine resin, Rink amide MBHA resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~1.1 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

  • Solid support for Fmoc-based solid phase peptide synthesis (SPPS) of peptide amides.
  • This solid support contains a modified Rink amide linker attached to the MBHA resin.
  • Cleavage of peptides is achieved by adding 95% TFA.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service