Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

802883

Sigma-Aldrich

Aldosterone-9,11,12,12-d4 solution

100 μg/mL in acetonitrile, ≥98 atom % D, ≥97% (CP)

Synonym(s):

T1AM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D4H24O5
Molecular Weight:
364.47
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥97% (CP)

form

liquid

concentration

100 μg/mL in acetonitrile

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2(C)C(CCC3[C@]2([2H])[C@@]([2H])(O[C@@H]4O)C([2H])([2H])[C@@]54C3CC[C@@H]5C(CO)=O)=C1

InChI

1S/C21H28O5/c1-20-7-6-12(23)8-11(20)2-3-13-14-4-5-15(16(24)10-22)21(14)9-17(18(13)20)26-19(21)25/h8,13-15,17-19,22,25H,2-7,9-10H2,1H3/t13?,14?,15-,17+,18-,19?,20+,21-/m1/s1/i9D2,17D,18D

InChI key

QUQBHBRVKLEOEI-SYXUCPCUSA-N

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service