Skip to Content
MilliporeSigma

809586

11-Deoxycorticosterone-2,3,4-13C3 solution

100 μg/mL in methanol, ≥99 atom % 13C, ≥98% (CP)

Synonym(s):

Cortexone-13C3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13C3C18H30O3
Molecular Weight:
333.44
PubChem Substance ID:
UNSPSC Code:
12142200
NACRES:
NA.12
EC Number:
200-659-6
Isotopic purity:
≥99 atom % 13C
Assay:
≥98% (CP)
Mass shift:
M+3
Form:
solution
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

≥99 atom % 13C

Quality Segment

assay

≥98% (CP)

form

solution

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

C[C@]1(C[13C]2)C(CCC3C1CC[C@@]4(C)C3CCC4C(CO)=O)=[13C][13C]2=O

InChI

1S/C21H27O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h15-18,22H,3-6,8-10,12H2,1-2H3/t15?,16?,17?,18?,20-,21-/m0/s1/i7+1,11+1,14+1

InChI key

HWSBXBKHQXFKBT-RZDKJTFNSA-N

General description

Tracer for steroid hormone biosynthesis studies. Applied in research on mineralocorticoid metabolism and adrenal steroidogenesis. Available from glass-bottle sizes perfect for laboratory research or bulk quantities for manufacturing. Inquire for bulk pricing.

Features and Benefits

  • We offer extensive product and packaging customization with fast turn-around, GMP capabilities, in-house technical expertise, and complete quality documentation.
  • We provide unmatched reliability, flexibility, scalability, and support.
  • Our portfolio includes key products available in many grades and quantities to suit specific project requirements.
  • Our secure supply chain ensures reliable access to materials for our customers, regardless of location.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library