Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

791318

Sigma-Aldrich

L-Histidine-d3 (α-d1, imidazole-2,5-d2) hydrochloride monohydrate

97% (CP), 98 atom % D, ≤18% D (deuterated on β, β-d2 positions)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D3H6N3O2HClH2O
Molecular Weight:
212.65
UNSPSC Code:
12352101

isotopic purity

≤18% D (deuterated on β, β-d2 positions)
98 atom % D

Assay

97% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+3

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service