Skip to Content
MilliporeSigma

M6633

α-D-Mannose pentaacetate

Synonym(s):

1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H22O11
CAS Number:
Molecular Weight:
390.34
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant (ivory nut palm)

Quality Segment

assay

≥98% (TLC)

form

powder

optical activity

[α]25/D 55.0 to 55.8°, c = 2.2% (w/v) in chloroform

technique(s)

thin layer chromatography (TLC): suitable

solubility

chloroform: 100 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

InChI

1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3

InChI key

LPTITAGPBXDDGR-UHFFFAOYSA-N

General description

Mannose is a monosaccharide.

Application

α-D-Mannose pentaacetate was used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C−H insertion.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library