Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773239

Sigma-Aldrich

1,4-Phenylene dimethacrylate

>99.0%, contains ≤1,000 ppm MeHQ as inhibitor

Synonym(s):

(1,4-phenylene) ester, 1,4-Phenylene bismethacrylate, 2-Methyl-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O4
CAS Number:
Molecular Weight:
246.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

>99.0%

form

solid

contains

≤1,000 ppm MeHQ as inhibitor

mp

89-91 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Oc1ccc(OC(=O)C(C)=C)cc1

InChI

1S/C14H14O4/c1-9(2)13(15)17-11-5-7-12(8-6-11)18-14(16)10(3)4/h5-8H,1,3H2,2,4H3

InChI key

MDMKOESKPAVFJF-UHFFFAOYSA-N

Application

This dimethacrylate is useful as a somewhat rigid crosslinker. The small spacer size can create a high density of crosslinking.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service