Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496758

Sigma-Aldrich

Bis[2-(methacryloyloxy)ethyl] phosphate

Synonym(s):

2-Hydroxyethyl methacrylate phosphate, Bis(2-methacryloyloxyethyl) acid phosphate, Bis(2-methacryloyloxyethyl) hydrogen phosphate, Bis(methacryloyloxyethyl) hydrogen phosphate, Bis(methacryloyloxyethyl) phosphate, Bis[2-(methacryloyloxy)ethyl] phosphate, Di(2-methacryloyloxyethyl) phosphate, Di-2-methacryloyloxyethyl acid phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2CH2CH2O]2P(O)OH
CAS Number:
Molecular Weight:
322.25
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

refractive index

n20/D 1.47 (lit.)

bp

221 °C (lit.)

density

1.28 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCOP(O)(=O)OCCOC(=O)C(C)=C

InChI

1S/C12H19O8P/c1-9(2)11(13)17-5-7-19-21(15,16)20-8-6-18-12(14)10(3)4/h1,3,5-8H2,2,4H3,(H,15,16)

InChI key

NXBXJOWBDCQIHF-UHFFFAOYSA-N

General description

Bis[2-(methacryloyloxy)ethyl] phosphate (BMEP) is a hydrophilic monomer with two polymerizable methacrylate groups and a centrally placed phosphate group. In presence of an acid, it can undergo spontaneous polymerization with a high degree of conversion. It is widely used as a precursor in the synthesis of poly (BMEP), dental resins, hydrogels, and fire-retardant polymer composites.

Application

Bis[2-(methacryloyloxy)ethyl] phosphate (BMEP) can be used:
  • As a bifunctional crosslinker to synthesize superabsorbent polymers (SAPs) via free radical polymerization. These SAPs can be used for agricultural applications.
  • To prepare self-etching primer formulations for enamel and dentine. Primers with BMEP exhibit good wetting, etching, and penetration in both dentine and enamel.
  • As a precursor to synthesize fire retardant nanocomposites via suspension polymerization.

Features and Benefits

  • Presence of dimethacrylate groups enables cross-linking and improves copolymerization.
  • Hydrophilicity and short carbon chains can enhance wetting.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Pinaeva et al.
Scientific reports, 10(1), 5776-5776 (2020-04-03)
Sorption mechanism of uranyl by poly(bis[2-(methacryloyloxy)ethyl] phosphate) (PB2MP) functionalised polyvinylidene fluoride (PVDF) track-etched membranes, PB2MP-g-PVDF, was investigated. It was found that uranyl sorption obeyed Langmuir isotherm model giving a maximum U(VI) membrane uptake of 6.73 μmol g-1 and an affinity
Ismail Anil et al.
Nanomaterials (Basel, Switzerland), 10(1) (2020-01-16)
The development of adsorbents with high adsorption capacity and fast separation is of utmost importance for the environmental management of dye-bearing wastewaters. Within this scope, crosslinked hydrogels including poly(vinylphosphonic acid) (PVPA) and bis[2-(methacryloyloxy)ethyl] phosphate (BMEP) were designed with varying mole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service