Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772623

Sigma-Aldrich

4,4,4-Trifluoro-1-butanol

98%

Synonym(s):

4,4,4-Trifluoro-n-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7F3O
CAS Number:
Molecular Weight:
128.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.343

bp

123-127 °C/760 mmHg

density

1.212 g/mL at 25 °C

functional group

fluoro
hydroxyl

SMILES string

OCCCC(F)(F)F

InChI

1S/C4H7F3O/c5-4(6,7)2-1-3-8/h8H,1-3H2

InChI key

VKRFUGHXKNNIJO-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.1 °F

Flash Point(C)

50.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service