Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769797

Sigma-Aldrich

Resomer® RG 755 S, Poly(D,L-lactide-co-glycolide)

ester terminated

Synonym(s):

Resomer® RG 755 S, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[C2H4O2]n[C2H2O2]mCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

amorphous

feed ratio

lactide:glycolide 75:25

viscosity

0.50-0.70 dL/g

transition temp

Tg 48-54 °C

storage temp.

2-8°C

SMILES string

O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Resomer® RG 755 S, Poly(D,L-lactide-co-glycolide), a biodegradable and biocompatible aliphatic polyester, can be used in the development of nano and microparticles. It is a pharmaceutical polymer, which finds potential usage in drug delivery and gene delivery. It forms a three-dimensional porous scaffold, which can be used in tissue engineering.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and evaluation of novel biodegradable microspheres based on poly (d, l-lactide-co-glycolide) and poly (varepsilon-caprolactone) for controlled delivery of doxycycline in the treatment of human periodontal pocket: in vitro and in vivo studies
Mundargi RC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 119(1), 59-68 (2007)
Nano/micro technologies for delivering macromolecular therapeutics using poly (D, L-lactide-co-glycolide) and its derivatives
Mundargi RC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 125(3), 193-209 (2008)
In vitro degradation of three-dimensional porous poly (D, L-lactide-co-glycolide) scaffolds for tissue engineering
Wu L and Ding J
Biomaterials, 25(27), 5821-5830 (2004)
Nanoparticles made of fluorescence-labelled poly (L-lactide-co-glycolide): preparation, stability, and biocompatibility
Weiss B, et al.
Journal of Nanoscience and Nanotechnology, 6(9-10), 3048-3056 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service