Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769789

Sigma-Aldrich

Resomer® RG 757 S, Poly(D,L-lactide-co-glycolide)

ester terminated, Lactide: Glycolide 75:25, lactide:glycolide 75:25

Synonym(s):

Resomer® RG 757 S, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[C2H4O2]n[C2H2O2]mCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25

Quality Level

form

amorphous

feed ratio

lactide:glycolide 75:25

viscosity

0.9-1.3 dL/g

storage temp.

2-8°C

SMILES string

O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Biodegradable, biocompatible, and bioresorbable copolymer composed of D,L-Lactide and Glycolide. This amorphous material has been used in sustained release drug delivery applications, by improving targeting and offering precise delivery timing. Degradation of this material has been thoroughly studied and has been shown to be safely resorbed by the body after implantation or injection. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service