Skip to Content
MilliporeSigma
All Photos(1)

Documents

764957

Sigma-Aldrich

Dipyrazino[2,3-f:2′,3′-h]quinoxaline-2,3,6,7,10,11-hexacarbonitrile

95% (HPLC)

Synonym(s):

1,4,5,8,9,11-Hexaazatriphenylenehexacarbonitrile, HAT-CN6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18N12
CAS Number:
Molecular Weight:
384.27
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95% (HPLC)

form

powder or crystals

mp

>500 °C

SMILES string

N#CC1=NC2=C(N=C1C#N)C3=C(N=C(C#N)C(C#N)=N3)C4=C2N=C(C#N)C(C#N)=N4

InChI

1S/C18N12/c19-1-7-8(2-20)26-14-13(25-7)15-17(29-10(4-22)9(3-21)27-15)18-16(14)28-11(5-23)12(6-24)30-18

InChI key

DKHNGUNXLDCATP-UHFFFAOYSA-N

Application

Hole transport layer materials for low voltage top-emitting organic green light emitting devices; a templating material that forces the copper phthalocyanine (CuPc) donor molecule to assume a vertical-standing morphology.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiangyong Pan et al.
Nanoscale, 10(2), 592-602 (2017-12-14)
We report an inverted and multilayer quantum dot light emitting diode (QLED) which boosts high efficiency by tuning the energy band alignment between charge transport and light emitting layers. The electron transport layer (ETL) was ZnO nanoparticles (NPs) with an
Synt. Metals, 162, 402-405 (2012)
Applied Physics Letters, 99, 043308/1-043308/3 (2011)
Zhen Bao et al.
Nanoscale, 12(21), 11556-11561 (2020-05-21)
Cadmium-free quantum dots (QDs) are attracting considerable research attention because of their low toxicity. However, the bandgap of most cadmium-free QDs avoids the pure-blue region, which leads to difficulty in realizing pure-blue quantum-dot light-emitting diodes (QLEDs). In this work, we

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service