Skip to Content
MilliporeSigma
All Photos(3)

Documents

776327

Sigma-Aldrich

4,4′,4′′-Tris[phenyl(m-tolyl)amino]triphenylamine

98.0%

Synonym(s):

4,4′,4′′-Tris[(3-methylphenyl)phenylamino]triphenylamine, N1-(3-methylphenyl)-N4,N4-bis[4-[(3-methylphenyl)phenylamino]phenyl]-N1-phenyl-1,4-benzenediamine, m-MTDATA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H48N4
CAS Number:
Molecular Weight:
789.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98.0%

form

powder

mp

194-203 °C
204-206 °C (lit.)

SMILES string

Cc1cccc(c1)N(c2ccccc2)c3ccc(cc3)N(c4ccc(cc4)N(c5ccccc5)c6cccc(C)c6)c7ccc(cc7)N(c8ccccc8)c9cccc(C)c9

InChI

1S/C57H48N4/c1-43-16-13-25-55(40-43)59(46-19-7-4-8-20-46)52-34-28-49(29-35-52)58(50-30-36-53(37-31-50)60(47-21-9-5-10-22-47)56-26-14-17-44(2)41-56)51-32-38-54(39-33-51)61(48-23-11-6-12-24-48)57-27-15-18-45(3)42-57/h4-42H,1-3H3

InChI key

DIVZFUBWFAOMCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hole-transporting layer and Hole Injection layer material for High-Performance OLEDs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rui Chen et al.
ChemSusChem, 11(9), 1467-1473 (2018-04-08)
Inverted perovskite solar cells (PSCs) with a p-i-n structure have attracted great attention. Normally, inorganic p-type metal oxides or polymers are used as the hole-transport material (HTM), a vital component in the inverted PSCs. However, this type of HTM often
A Novel Family of Boron-Containing Hole-Blocking Amorphous Molecular Materials for Blue- and Blue?Violet-Emitting Organic Electroluminescent Devices
Kinoshita, M.; et al.
Advances in Functional Materials, 12, 780-786 (2002)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service