Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764388

Sigma-Aldrich

Tetraethylene glycol p-toluenesulfonate

97%

Synonym(s):

2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate, 2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethyl toluene-4-sulfonate, 2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethyl toluenesulfonate, 2-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]ethyl tosylate, Monotosylated tetraethylene glycol, Tetraethylene glycol monotosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O7S
CAS Number:
Molecular Weight:
348.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.507

density

1.202 g/mL at 25 °C

functional group

ether
hydroxyl
tosylate

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOCCOCCOCCO

InChI

1S/C15H24O7S/c1-14-2-4-15(5-3-14)23(17,18)22-13-12-21-11-10-20-9-8-19-7-6-16/h2-5,16H,6-13H2,1H3

InChI key

FGDJUEZBMFELRW-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service