Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

705276

Sigma-Aldrich

Tetraethyleneglycol monomethyl ether

technical grade, average MN 200, methoxy, hydroxyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20O5
CAS Number:
Molecular Weight:
208.25
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Tetraethyleneglycol monomethyl ether, technical grade

grade

technical grade

Quality Level

form

liquid

mol wt

average Mn 200

refractive index

n20/D 1.444

density

1.045 g/mL at 25 °C

Ω-end

hydroxyl

α-end

methoxy

SMILES string

COCCOCCOCCOCCO

InChI

1S/C9H20O5/c1-11-4-5-13-8-9-14-7-6-12-3-2-10/h10H,2-9H2,1H3

InChI key

ZNYRFEPBTVGZDN-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel Martínez-Edo et al.
Nanomaterials (Basel, Switzerland), 9(9) (2019-09-01)
A straightforward methodology for the synthesis of isothiocyanate-functionalized mesoporous silica nanoparticles (MSNs) by exposure of aminated MSNs to 1,1'-thiocarbonyldi-2(1H)-pyridone is reported. These nanoparticles are chemically stable, water tolerant, and readily react with primary amines without the formation of any by-product.
Tian Zhou et al.
Nature communications, 7, 11119-11119 (2016-03-25)
Precise synthesis of polymer brushes to modify the surface of nanoparticles and nanodevices for targeted applications has been one of the major focuses in the community for decades. Here we report a self-assembly-assisted-grafting-to approach to synthesize polymer brushes on flat
Shota Konno et al.
PloS one, 13(8), e0202816-e0202816 (2018-08-28)
As a frontier topic of soft condensed matter physics, irreversible aggregation has drawn attention for a better understanding of the complex behavior of biomaterials. In this study, we have described the synthesis of an artificial amphiphilic molecule, an alternating tetra-block-like

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service