Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

763314

Sigma-Aldrich

sSPhos Pd G2

95%

Synonym(s):

Chloro(sodium-2-dicyclohexylphosphino-2′,6′-dimethoxy-1,1′-biphenyl-3′-sulfonate)[2-(2′-amino-1,1′-biphenyl)]palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H44ClNNaO5PPdS
CAS Number:
Molecular Weight:
822.66
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

193-194 °C (decomposition)

functional group

phosphine

SMILES string

[Na+].Nc1ccccc1-c2ccccc2[Pd]Cl.COc3ccc(c(OC)c3-c4ccccc4P(C5CCCCC5)C6CCCCC6)S([O-])(=O)=O

InChI

1S/C26H35O5PS.C12H10N.ClH.Na.Pd/c1-30-22-17-18-24(33(27,28)29)26(31-2)25(22)21-15-9-10-16-23(21)32(19-11-5-3-6-12-19)20-13-7-4-8-14-20;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;;/h9-10,15-20H,3-8,11-14H2,1-2H3,(H,27,28,29);1-6,8-9H,13H2;1H;;/q;;;2*+1/p-2

InChI key

MWQYWXJKMDRSJN-UHFFFAOYSA-L

Application

sSPhos Pd G2 can be used as a water-soluble Pd-precatalyst in:
  • DNA-compatible Suzuki-Miyaura reaction between DNA-linked aryl halides and various boronic acids/esters including heteroaryl boronates.
  • DNA-compatible hydroxycarbonylation reactions.
  • The cross-coupling reaction between boronic ester and brominated cyclic nucleotide to synthesize cyclic adenosine and guanosine monophosphate in the presence of potassium phosphate and hydrochloric acid.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Yuan Li et al.
Bioconjugate chemistry, 29(11), 3841-3846 (2018-10-20)
DNA-encoded chemical libraries (DELs) are a cost-effective technology for the discovery of novel chemical probes and drug candidates. A major limiting factor in assembling productive DELs is the availability of DNA-compatible chemical reactions in aqueous media. In an effort to
Wentao Liu et al.
Bioorganic & medicinal chemistry, 38, 116118-116118 (2021-04-12)
A robust palladium-catalyzed hydroxycarbonylation of aryl halides on DNA has been developed. Instead of Mo(CO)6 as a source of carbon monoxide as previously described in the literature, chloroform was used as a surrogate in this report for the purpose of
Marco Lelle et al.
Chembiochem : a European journal of chemical biology, 21(16), 2311-2320 (2020-04-01)
High-affinity fluorescent derivatives of cyclic adenosine and guanosine monophosphate are powerful tools for investigating their natural targets. Cyclic nucleotide-regulated ion channels belong to these targets and are vital for many signal transduction processes, such as vision and olfaction. The relation

Articles

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service