Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

761990

Sigma-Aldrich

(R)-1-Methyl-2-pyrrolidinemethanol

97%

Synonym(s):

(R)-2-Hydroxymethyl-1-methylpyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 4.469

bp

68-71 °C (lit.)

density

0.971 g/mL at 25 °C

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

OC[C@@H]1N(C)CCC1

InChI

1S/C6H13NO/c1-7-4-2-3-6(7)5-8/h6,8H,2-5H2,1H3/t6-/m1/s1

InChI key

VCOJPHPOVDIRJK-ZCFIWIBFSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

160.0 °F

Flash Point(C)

71.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service