Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748773

Sigma-Aldrich

Ethyl 1H-imidazole-1-carboxylate

Synonym(s):

Heller-Sarpong Reagent, N-Carboethoxyimidazole, EtImC, Ethyl imidazolecarbamate, Sarpong reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.472

density

1.162 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)n1ccnc1

InChI

1S/C6H8N2O2/c1-2-10-6(9)8-4-3-7-5-8/h3-5H,2H2,1H3

InChI key

YICRPEGTQDSFEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.9 °F

Flash Point(C)

104.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service