Skip to Content
MilliporeSigma
All Photos(1)

Documents

662933

Sigma-Aldrich

6-Methoxy-2-pyridinecarboxaldehyde

97%

Synonym(s):

2-Formyl-6-methoxypyridine, 6-Methoxypyridine-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.532

bp

103-104 °C/20 mmHg

density

1.140 g/mL at 25 °C

SMILES string

[H]C(=O)c1cccc(OC)n1

InChI

1S/C7H7NO2/c1-10-7-4-2-3-6(5-9)8-7/h2-5H,1H3

InChI key

YDNWTNODZDSPNZ-UHFFFAOYSA-N

Application

Synthesis of the corresponding α-pyridoin via treatment with NaCN.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Hatanaka et al.
Bioorganic & medicinal chemistry, 13(24), 6763-6770 (2005-08-30)
Focusing on alpha-pyridoin (1, 1,2-di(2-pyridyl)-1,2-ethenediol) as the lead compound of the novel antioxidative enediol, we synthesized 5,5'- or 6,6'-bis-substituted derivatives of 1 from disubstituted pyridines. The antioxidant activity of 1 and its synthetic derivatives 2-7 was evaluated by DPPH (1,1-diphenyl-2-picrylhydrazyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service