Skip to Content
MilliporeSigma
All Photos(1)

Documents

744646

Sigma-Aldrich

(S)-(+)-N-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)-dibenzo[b,f]azepine

≥95% (elemental analysis)

Synonym(s):

5-(11bS)-Dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-yl-5H-dibenz[b,f]azepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H22NO2P
CAS Number:
Molecular Weight:
507.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (elemental analysis)

form

powder

composition

carbon content, 76.4-84.5 wt. %

SMILES string

C1(C=CC=C2)=C2C(C3=C(C=CC=C4)C4=CC=C3OP5N6C7=C(C=CC=C7)C=CC8=C6C=CC=C8)=C(O5)C=C1

InChI

1S/C34H22NO2P/c1-5-13-27-23(9-1)19-21-31-33(27)34-28-14-6-2-10-24(28)20-22-32(34)37-38(36-31)35-29-15-7-3-11-25(29)17-18-26-12-4-8-16-30(26)35/h1-22H

InChI key

KOVOHCVLXRKTGN-UHFFFAOYSA-N

Application

Phosphorous precursor for ligand synthesis

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drinkel, E., et al.
Organometallics, 29, 2503-2514 (2010)
Mariz, R., et al.
Organometallics, 27, 6605-6613 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service