Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665460

Sigma-Aldrich

(3aR,8aR)-(−)-(2,2-Dimethyl-4,4,8,8-tetraphenyl-tetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34NO4P
CAS Number:
Molecular Weight:
539.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

218-221 °C

functional group

amine
ether
ketal
phenyl

SMILES string

CN(C)P1OC([C@@H]2OC(C)(C)O[C@H]2C(O1)(c3ccccc3)c4ccccc4)(c5ccccc5)c6ccccc6

InChI

1S/C33H34NO4P/c1-31(2)35-29-30(36-31)33(27-21-13-7-14-22-27,28-23-15-8-16-24-28)38-39(34(3)4)37-32(29,25-17-9-5-10-18-25)26-19-11-6-12-20-26/h5-24,29-30H,1-4H3/t29-,30-/m1/s1

InChI key

DRPNADVMTMEWQY-LOYHVIPDSA-N

Application

Phosphoramidite ligand employed in an asymmetric, iridium-catalyzed, allylic allylation of propenylboronates. Also used in a copper-catalyzed asymmetric conjugate addition of diethylzinc to α,β-unsaturated imines derived from amino acids.

Legal Information

Sold under license from DSM for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron Letters, 48, 3305-3305 (2007)
Francisco Palacios et al.
Organic letters, 8(23), 5405-5408 (2006-11-03)
[Structure: see text] A highly enantioselective synthesis of alpha-dehydroaminoacids with a stereogenic center in the gamma position through copper-catalyzed asymmetric conjugate addition of diethylzinc to alpha,beta-unsaturated imines using a TADDOL-derived phosphoramidite complex is reported.

Articles

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service