Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741345

Sigma-Aldrich

2-Chloro-5-(difluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClF2N
CAS Number:
Molecular Weight:
163.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.492

density

1.368 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

FC(F)c1ccc(Cl)nc1

InChI

1S/C6H4ClF2N/c7-5-2-1-4(3-10-5)6(8)9/h1-3,6H

InChI key

OSQYJPKSEWBNOE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

225.0 °F

Flash Point(C)

107.22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service