Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

720313

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)pyrimidine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2ClF3N2
CAS Number:
Molecular Weight:
182.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

48-52 °C

storage temp.

−20°C

SMILES string

FC(F)(F)c1cnc(Cl)nc1

InChI

1S/C5H2ClF3N2/c6-4-10-1-3(2-11-4)5(7,8)9/h1-2H

InChI key

TYCYTQLXAIDJNF-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

160.0 °F - closed cup

Flash Point(C)

71.1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service